Imprimer
Millicell® Cell Culture Inserts, Hydrophilic PTFE (Biopore™) Membrane
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PICM0RG50 PICM01250 PICM03050
MILFPICM0RG50EA 562 EUR
MILFPICM0RG50 MILFPICM01250 MILFPICM03050
Millicell® Cell Culture Inserts, Hydrophilic PTFE (Biopore™) Membrane
Microplaques Inserts pour plaques de cultures
With Millicell inserts, attachment or suspension cells can access media from both their apical and basolateral sides. Cell growth, structure, and function more closely mimic what occurs in vivo. In addition, Millicell inserts make it possible to study both sides of the cell monolayer.
The inserts are easily prepared for SEM and TEM visualising techniques, and they are compatible with cellular and/or fluorescent stains.