Imprimer
Millex® Automation-Compatible Filter Units, with Durapore (PVDF) Membrane
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SLHVBZ5NZ SLHVDZ5NZ SLHVBZ5NK SLHVDZ5NK
MILFSLHVBZ5NZEA 827 EUR
MILFSLHVBZ5NZ MILFSLHVDZ5NZ MILFSLHVBZ5NK MILFSLHVDZ5NK
Millex® Automation-Compatible Filter Units, with Durapore (PVDF) Membrane
Filtres Filtres-seringue
Automation compatible filter units are specially designed to provide efficient filtration for dissolution and other automated sample preparation applications.
  • Domed housing ensures reliable delivery of filters
  • Pressure-resistant housing resists bursting
  • Luer-Lok® connection optimised for precise alignment and fit
Automation compatible filter units are available in a variety of membrane types and pore sizes, including units with pre-filters for particulate-laden samples. Used in the process of drug dissolution testing, automated sample preparation, clarification of solutions containing high particle levels etc.
Max. temperature: 45 °C
Applications: drug dissolution testing, automated sample preparation, clarification of solutions containing high particle levels.