Imprimer
Steriflip® Bottle Top Vacuum Filtration Systems, Durapore® (PVDF) membrane
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SE1M003M00 SE1M179M6
MILFSE1M003M00EA 254 EUR
MILFSE1M003M00 MILXSE1M179M6
Steriflip® Bottle Top Vacuum Filtration Systems, Durapore® (PVDF) membrane
Filtres Unités de filtration sous vide
The Steriflip® filter unit performs vacuum assisted, closed system ensures sample sterility while enabling fast separation of large volumes of cellular material for efficient recoveries. It also eliminates messy pouring during sample transfer. Filters come in different pore sizes that are ideal for cell isolation.
  • Inlet fittings are double lead thread with vaccum port
  • Attaches to any standard 50 ml centrifuge tube, eliminating unnecesary materials
Max. temperature: 45 °C