Imprimer
Millex® Syringe Filters, MCE Membrane
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SLGS033SS SLGSV255F SLHA033SS SLGL0250S SLAA033SS SLGS033SB SLHA033SB SLAA033SB SLAAV255F SLHAM33SS SLGSM33SS
MILFSLGS033SSEA 237 EUR
MILFSLGS033SS MILFSLGSV255F MILFSLHA033SS MILFSLGL0250S MILFSLAA033SS MILFSLGS033SB MILFSLHA033SB MILFSLAA033SB MILFSLAAV255F MILFSLHAM33SS MILFSLGSM33SS
Millex® Syringe Filters, MCE Membrane
Filters Syringe Filters
Filters for sterilising or clarifying biological solutions. The larger filtration area increases flow rate and throughput. It also makes it easier to filter solutions because it reduces the pressure required to empty the syringe.
Manufacturing occurs in a controlled environment using an automated process. Provided with a certificate of quality.