Imprimer
Millicell® EZ Slides
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PEZGS0896 PEZGS0496 PEZGS0416 PEZGS0816
MILFPEZGS0896EA 967 EUR
MILFPEZGS0896 MILFPEZGS0496 MILFPEZGS0416 MILFPEZGS0816
Millicell® EZ Slides
Lames Lames de microscopes
Millicell® EZ Slide allows the user to grow, fix, and stain cells directly on a microscope slide. There is no need to disassemble the unit or transfer cells from one device to another prior to analysis.
The polypropylene wells are resistant to most fixatives used, so the cells can be grown on the slide and stained right in the well. Once stained, the microscope slide can be easily removed by pressing down the four side tabs of the unit and lifting the wells off of the slide.
This easy slide removal reduces the risk of damaging the cells and no special tools are required to remove the wells from the slide. Raised corner ridges on the cover facilitate stacking of multiple units in the incubator, or they can be incubated in the tray supplied with the units.
Raised corner ridges on the lid facilitate stacking of multiple units.
Techniques: Cell based assay, suitable cell culture, mammalian: suitable immunocytochemistry: suitable.