Imprimer
Chemical Duty Pumps
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
WP6110060 WP6111560
MILFWP6110060EA 1300 EUR
MILFWP6110060 MILFWP6111560
Chemical Duty Pumps
Pompe Pompes à vide
The Chemical Duty Pump has a chemically-resistant head and diaphragm for use with corrosive chemicals and solvents.
  • High flow rates decrease filtration process time
  • Optional chemical-resistant internal components for compatibility with corrosive vapours
  • Portable design allows for easy sharing between workstations
Pumps come with 70 cm of ¹/₄" tubing and a Millex FA50 filter for in-line moisture protection. UL listed and CE marked. Chemical Duty Pump has a chemically-resistant head and diaphragm for use with corrosive chemicals and solvents.
Applications:
Vacuum source for filter holders, manifolds, and devices, including Samplicity® filtration system, SNAP i.d.® 2.0 system, Multiscreen®HTS filtration manifold, and Stericup® filters