Imprimer
SNAP i.d.® Starter Packages
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
STARTMIDI STARTMINI STARTMM STARTMULTI
MILFSTARTMIDIEA 2120 EUR
MILFSTARTMIDI MILFSTARTMINI MILFSTARTMM MILFSTARTMULTI
SNAP i.d.® Starter Packages
Analyseurs Analyseur de protéines
Western blot packages with unbelievable savings! Save time and money by choosing the package that best serves your needs! The SNAP i.d.® system applies a vacuum to actively drive reagents through the membrane, promoting antigen binding and thorough washing, enabling you to better optimise your Western blotting.
  • Superior: Increased antibody-antigen binding, enhanced washes, and antibody recollection
  • Fast: 30-minute immunodetection
  • Faster testing of different antibodies
  • Higher throughput of Western blots each day
  • Draws reagents through blotting membrane
  • Minimises over-blocking
  • Thoroughly flushes membranes instead of just rinsing
  • Reduces incubation times
Flexible: Two gel sizes, mini (7,5×8,4 cm) and midi (8,7×13,5 cm).
Informations de livraison: SNAP i.d.™ 2.0 Protein Detection system, Immobilon™-P Membrane, 0,45 μm, 26,5 cm×3,75 m roll, SNAP i.d.™ 2.0 Blot Holders, 100 units per pack, Bløk™-CH Noise Cancelling Reagent, Luminata™ Classico Western HRP Substrate, 100 ml, Reblot™ Plus Strong Antibody Stripping Solution, 10X, 50 ml.