Imprimer
MultiScreen® PCR 96 Filter Plates
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MSNU03050 MSNU03010
MILFMSNU03050EA 0 EUR
MILFMSNU03050 MILFMSNU03010
MultiScreen® PCR 96 Filter Plates
Microplaques Plaques de filtration
The polymerase chain reaction (PCR) is a highly effective fundamental technique in molecular biology. It offers a fast and efficient way to amplify specific DNA or RNA sequences from nucleic acids derived from different sources using enzymatic processes in a laboratory setting.
  • Application: Sample preparation
  • Binding type: Low binding surface
  • Ambient
We have an extensive range of PCR consumables and equipment designed to facilitate every stage of your PCR workflow MultiScreen-PCR 96 Filter plates can be used to purify PCR products to be used in further downstream applications.
MultiScreen-PCR 96 filter plate is suitable for use:
-In receptor/ligand binding assays
-In sample preparation
-In enzyme assays with precipitation
-In DNA purification
-In polymerase chain reaction (PCR)
-In purifying polymerase chain reaction (PCR) products for mutation analysis
-In purifying PCR products for measuring CAG repeats
-In purifying PCR products for sequencing