Imprimer
Millex® Samplicity Syringe Filters
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SAMP2LCRB SAMP2LGNB SAMP2LCRK SAMP2LGNK
MILFSAMP2LCRBEA 1130 EUR
MILFSAMP2LCRB MILFSAMP2LGNB MILFSAMP2LCRK MILFSAMP2LGNK
Millex® Samplicity Syringe Filters
Filtres Filtres-seringue
Millex® Filters for the Samplicity® G2 System
  • Samplicity® G2 filtration system enables vacuum driven filtration of up to 8 samples at once, directly into HPLC vials
  • Millex® Filters for the Samplicity® G2 system provide adaptor funnels and Millex® 33 mm filters
  • Samplicity® G2 system has lower ergonomic impact than manual syringe filtration
  • Available in 0,20 µm and 0,45 µm pore sizes with hydrophilic PTFE, Durapore® PVDF, Millipore Express® PLUS PES, or nylon membranes
Applications: HPLC; filtration of viscous or particulate laden samples for HPLC.