Imprimer
Strat-M® PES Membrane Filters
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SKBM02560 SKBM04760
MILFSKBM02560EA 924 EUR
MILFSKBM02560 MILFSKBM04760
Strat-M® PES Membrane Filters
Filtres Filtres à membrane
Strat-M membrane for transdermal diffusion testing, experience the unmatched predictability of Strat-M membrane - a synthetic, non-animal based model for transdermal diffusion testing that is predictive of diffusion in human skin without lot-to-lot variability, safety and storage limitations.
  • Active pharmaceutical ingredients (API)
  • Cosmetic actives
  • Formulations
  • Personal care products
  • Pesticides and chemicals
  • Thickness: 300 μm
  • Ambient
Human skin correlation:
Strat-M membrane is predictive of diffusion in human skin for a wide range of compounds and formulations. Use the Compound Correlation Tool to learn how well Strat-M membrane will work for you.
Reproducible diffusion data:
No need to repeat testing each time a new lot of skin arrives in the lab. Strat-M membrane is a highly consistent, engineered material that allows you to compare today’s diffusion data with data you will generate next week, or next month. Furthermore, lot-to-lot consistency brings a new level of clarity to your test data, simplifying your analysis.
Sensitive to enhancers:
Strat-M membrane exhibits differential permeability in the presence of enhancers and is compatible with common formulation formats, so it is appropriate for use during formulation optimisation.
Ultimate experimental convenience:
Strat-M membrane was designed with convenience in mind. The Strat-M membrane is shelf-stable and stocked in the warehouse so procurement and storage is a breeze. Each membrane is individually packaged as precut discs and do not need to be hydrated prior to use. Simply remove the Strat-M membrane disc from its pouch and load into a Franz cell.
Application:
Strat-M membrane is a synthetic, non-animal based model for transdermal diffusion testing that is predictive of diffusion in human skin without lot-to-lot variability, safety and storage limitations.