Imprimer
Multiscreen® 96-Well Plates with Polycarbonate Membrane
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MAMIC3S10 MAMIC8S10 MAMIC5S10
MILFMAMIC3S10EA 1140 EUR
MILFMAMIC3S10 MILFMAMIC8S10 MILFMAMIC5S10
Multiscreen® 96-Well Plates with Polycarbonate Membrane
Microplaques Plaques de filtration
HTS filter plates (Plate catalogue numbers that start with MS): MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters. Filter plates are available in 96- and 384-well platforms with a broad selection of membranes and plastics. Millipore's new MultiScreen HTS+ Hi flow filter plate design lessens non-specific binding and reduces variability in both background and signal intensities specifically for biochemical screening assays.
Classic filter plates (Plate catalogue numbers that start with MA): The original multiwell design is valuable for many applications which require immersing the filter in an assay tray. The easily removable underdrain and unique plate design are ideal for assays such as Elispot, PAMPA, cell screening (migration, invasion or chemotaxis) or for topcount detection users.
Millicell assay plates: In addition to the traditional assay plate format, filter plates are also available with cell culture trays for growing and assaying suspension and adherent cell lines. Cells grown on filter plates differentiate better than those grown on traditional plastic substrates, resulting in more in vivo-like results This filter plate/tray combination can be used for a variety of cellular assays, including, transport, toxicity and screening assays.
Informations sur les accessoires: The MultiScreenHTS vacuum manifold was designed specifically to work with MultiScreenHTS filter plates. The combination results in optimal filtration characteristics and seamless automation integration.