Imprimer
MultiScreenHTS Filter Plates with Hydrophilic Durapore® PVDF membrane
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MSBVN1250 MSBVN1210 MSDVN6B50 MSHVN4550 MSBVS1210 MSDVN6550 MSHVN4510 MSGVS2210 MSHVS4B10 MSGVN2250 MSGVN2B50 MSHVS4510 MZHVN0W50 MSDVN6510 MSBVN1B50 MZHVN0W10 MSGVN2210
MILFMSBVN1250EA 1500 EUR
MILFMSBVN1250 MILFMSBVN1210 MILFMSDVN6B50 MILFMSHVN4550 MILFMSBVS1210 MILFMSDVN6550 MILFMSHVN4510 MILFMSGVS2210 MILFMSHVS4B10 MILFMSGVN2250 MILFMSGVN2B50 MILFMSHVS4510 MILFMZHVN0W50 MILFMSDVN6510 MILFMSBVN1B50 MILFMZHVN0W10 MILFMSGVN2210
MultiScreenHTS Filter Plates with Hydrophilic Durapore® PVDF membrane
Microplaques Plaques de filtration
MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters. Filter plates are available in 96- and 384-well platforms with a broad selection of membranes and plastics.
  • Hydrophilic Durapore® polyvinylidene fluoride (PVDF) membrane
Millipore's new MultiScreen HTS+ Hi flow filter plate design lessens non-En savoir plus