Imprimer
Stericup® E-GP Funnel-less Sterile Vacuum Filtration System
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SEGPU0538 SEGPU0545 SEGPU1138 SEGPU1145
MILFSEGPU0538EA 238 EUR
MILFSEGPU0538 MILFSEGPU0545 MILFSEGPU1138 MILFSEGPU1145
Stericup® E-GP Funnel-less Sterile Vacuum Filtration System
Filtres Unités de filtration sous vide
Work with ease and filter with confidence with Stericup® and Steritop® filtration systems. The Stericup® E and Steritop® E sterile filters have been designed to reduce plastic and packaging.
  • The filter collar disconnects from the receiver bottle with just a quarter turn reducing the likelihood of spillage and making it easier to manipulate on the bench or under the hood
  • Bold product identification on the filter collar plainly displays membrane characteristics, including composition and pore size, for rapid, application-appropriate product selection
  • Click seal cap is flanged and modified to enable a firm grip in wet or dry conditions and a tactile stop confirms secure closure. Worry less about the possibility of spilling or contamination of sterilised contents
  • Frosted, ample writing surface on the receiver bottle and lighter cap color facilitate clear labelling to improve legibility and reduce unreadable smears
  • Click seal cap rests on the side to avoid risk of contamination
  • Receiver bottles are stackable to save storage space
This Stericup® E is a 38 mm thread collar which uses a 0,22 μm pore size polyethersulfone (PES) membrane filter unit combined with a 500, 1000 ml receiver flask and cap for processing and storage. The 38 mm thread directly connects to our media bottles and majority of other standard commercial media bottles.
This Stericup® E is a 45 mm thread collar which uses a 0,22 μm pore size polyethersulfone (PES) membrane filter unit combined with a 500, 1000 ml receiver flask and cap for processing and storage. The 45 mm thread directly connects to wider neck media bottles (such as Gibco® or glass bottles).
Attention: For research use only. Use only glass or plastic bottles designed for vacuum applications.