Imprimer
Microcon® DNA Fast Flow Centrifugal Filters
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MRCF0R100 MRCF0R100ET
MILFMRCF0R100EA 562 EUR
MILFMRCF0R100 MILFMRCF0R100ET
Microcon® DNA Fast Flow Centrifugal Filters
Filtres Filtres pour centrifugation
Optimized for the concentration and recovery of genomic DNA with SDS buffer. The low nonspecific binding characteristics of the membrane and the other device components, coupled with its medical-grade O-ring seal, allow the device to accommodate several wash steps with minimal sample loss.
  • High recovery for small volumes with reverse spin (concentration factor <20X)
  • Low-binding Ultracel® membrane
  • Fast processing
Sample applications:
Recovery of genomic DNA for forensic applications.
Concentration and desalting of nucleic acids (high recovery alternative to ethanol precipitation).
Removal of primers from amplified DNA.
Easy filtrate recovery, exploiting the flat membrane design.
Extract regular organic DNA to compare DNA yields from alternative extraction methods.
Simply and efficiently concentrate and desalt solutions of any macromolecule, using any centrifuge that can accept 1,5 ml tubes. With the low-binding Ultracel membrane, Microcon filters offer:
•Typical recoveries of greater than 95%, even for dilute solutions.
•Reverse spin to maximize recovery, even in the smallest samples.
•Convenient storage of filtrate or concentrated sample in standard microfuge tube.
•Concentration factors up to 100X.