Imprimer
Microcon® Centrifugal Filters
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MRCF0R030 MRCPRT010
MILFMRCF0R030EA 562 EUR
MILFMRCF0R030 MILFMRCPRT010
Microcon® Centrifugal Filters
Filtres Filtres pour centrifugation
Simply and efficiently concentrate and desalt solutions of any macromolecule with the low-binding Ultracel® membrane, using any centrifuge that can accept 1,5 ml tubes.
  • Dual-cycle EtO treatment on the Microcon® PCR Grade Filter has been shown to render contaminating DNA unamplifiable
  • Typical recoveries of >95%, even for dilute solutions
  • Reverse spin to maximize recovery, even in the smallest samples
  • Convenient storage of filtrate or concentrated sample in standard microfuge tube
  • Concentration factors up to 100X