Imprimer
Orion™ Conductivity Standards and Solutions, Thermo Scientific
Marques : Thermo Orion
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
990106 011001 011006 01100910 11007 11008 O1100510 O1100710
663-0044EA 103 EUR
663-0044 ORIO011001 663-0022 ORIO01100910 ORIO11007 ORIO11008 ORIOO1100510 ORIOO1100710
Orion™ Conductivity Standards and Solutions, Thermo Scientific
Étalons Étalons pour la mesure de conductivité
Perform virtually any electrochemistry analysis with these conductivity standards and solutions. All conductivity standards are traceable to NIST™ standard reference materials.
  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
  • Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature
  • For use with Thermo Scientific™ Orion™ Conductivity Probes
Ensure optimal conductivity probe maintenance and performance with Thermo Scientific™ Orion™ Conductivity Standards and Solutions. Conductivity conditioning solutions, conductivity standards and TDS standards are available for use with your Orion™ 2 Cell Conductivity Probe or Orion™ DuraProbe™ 4 Cell Lab and Field Conductivity Probes. Use these solutions during calibration, cleaning and storage of your probes to help eliminate build-up on the conductivity electrode cell or bulb surface and to attain accurate, consistent readings.