Imprimer
Millex® MCE Syringe Filters
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SLAA025NB SLHA025NB SLGS025NB SLHA02510 SLAA025NK SLGS02510
MILFSLAA025NBEA 671 EUR
MILFSLAA025NB 516-6552 MILFSLGS025NB MILFSLHA02510 MILFSLAA025NK MILFSLGS02510
Millex® MCE Syringe Filters
Filtres Filtres-seringue
Filters for sterilising or clarifying biological solutions. The larger filtration area increases flow rate and throughput. It also makes it easier to filter solutions because it reduces the pressure required to empty the syringe.
  • Hydrophilic mixed cellulose esters (MCE) membrane available in 0,22 µm, 0,45 µm and 0,80 µm pore sizes
  • Housing is made from a low-extractable polyvinylidene chloride (PVC)
  • Holdup volume<100 μl
  • Hydrophilic
Applications: Clarification of aqueous solutions, general laboratory filtration.