Imprimer
Millex® SLHPX13 Syringe Filters, Polyethersulfone
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SLHPX13NK SLHPX13NL
MILFSLHPX13NKEA 2020 EUR
MILFSLHPX13NK MILFSLHPX13NL
Millex® SLHPX13 Syringe Filters, Polyethersulfone
Filtres Filtres-seringue
Syringe filters are single-use, membrane-based devices used for the removal of particulate impurities from liquid and gas samples prior to analysis by methods such as HPLC, ion chromatography, gas chromatography, ICP, and dissolution testing. Proper filtration of samples improves the quality and consistency of analytical results and decreases instrument downtime.
  • Non-sterile Millex syringe filters with Millipore Express PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions
  • Available in 0,22 µm and 0,45 µm pore sizes and two diameters to suit your application needs
  • Low protein binding to minimize interaction with your sample and maximize recovery
  • Polypropylene housing, offering a low level of extractables and broad chemical compatibility
  • Holdup volume: ≤15 μl (after air purge) hydrophilic
  • Bubble point: ≥2,0 bar, air with water at 23 °C
  • Height: 21 mm
Disposable syringe filters are commonly used for fast and efficient filtering. Selection should be by application. Sterile syringe filters are used to sterilise solutions or clarify sterile solutions, while nonsterile syringe filters are used for general filtration and sample purification. Syringe filters are categorized by two essential characteristics - membrane and filter housing.
Both characteristics must be compatible with the application and solution being filtered. Syringe filter membranes are matched by composition, filter diameter, and pore size, while syringe filter housings are matched to end applications based on composition and format.
Techniques for SLHPX13: HPLC, UV/Vis spectroscopy, analytical sample preparation, fast flow filtration and low protein-binding filtration.
Parameter:
10 bar max. inlet pressure (10,34 min-1)
104 L/min flow rate
104 ml/min-cm² water flow rate
45 °C max. temp.