Imprimer
TableWorx Performance Work Tables, 304 Stainless Steel Work Surface
Fournisseur: Metro International
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TWM3060SI304S TWM3072SI304S TWM3096SI304S
METOTWM3060SI304SEA 0 EUR
METOTWM3060SI304S METOTWM3072SI304S METOTWM3096SI304S
TableWorx Performance Work Tables, 304 Stainless Steel Work Surface
Mobilier Tables
TableWorx is a truly modular platform of performance work tables designed to allow you to customise your perfect solution.
All stainless series TableWorx performance work tables feature a 16-gauge, type 304 stainless steel work surface reinforced with 16-gauge hat channels. The sound deadened work surface features 90° turned down edges on all four sides. All sizes are available with an optional 51 mm stainless steel backsplash, which is formed into the work surface.
All stainless series TableWorx performance work tables feature type 304 stainless steel legs and leg mounts. A sturdy, type 304 stainless steel I-frame is ideal for applications that require two-sided access to the work table. It can also be used as a foot rest for seated applications.
Mobile-ready versions are also available, and can accept any Metro 25 mm stem caster. Please note that casters are NOT included with mobile-ready TableWorx models and must be ordered separately.