Imprimer
Freezer Storage Boxes for 50 ml Centrifuge Tubes
Fournisseur: LABCON
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2704-220-000 2704-220-000-9
LBCN2704-220-000EA 123 EUR
LBCN2704-220-000 LBCN2704-220-000-9
Freezer Storage Boxes for 50 ml Centrifuge Tubes
Boîtes Boîtes de rangement
Labcon 16 or 36 Place Freezer Storage Boxes for 50 ml centrifuge tubes. These boxes are designed to hold 50 ml tubes and are perfect for long term storage of frozen samples.
  • Made of polypropylene
  • Freezable to −80 °C
  • Autoclavable
  • Includes clear lid
They include a white upper body that can be easily labeled with a lab marker, a numerical reference grid, and a pull tab to make removal from storage racks easier.
Informations de livraison: A clear cover is included.
Cdt: Two boxes per case.