Imprimer
ESD Safety Shoes, Lace-Up, uvex 1 x-craft planet, 68082
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6808.2/46 6808.2/44 6808.2/52 6808.2/35 6808.2/43 6808.2/50 6808.2/38 6808.2/37 6808.2/51 6808.2/48 6808.2/45 6808.2/36 6808.2/40 6808.2/41 6808.2/42 6808.2/49 6808.2/39 6808.2/47
UVEX6808.2/46EA 165 EUR
UVEX6808.2/46 UVEX6808.2/44 UVEX6808.2/52 UVEX6808.2/35 UVEX6808.2/43 UVEX6808.2/50 UVEX6808.2/38 UVEX6808.2/37 UVEX6808.2/51 UVEX6808.2/48 UVEX6808.2/45 UVEX6808.2/36 UVEX6808.2/40 UVEX6808.2/41 UVEX6808.2/42 UVEX6808.2/49 UVEX6808.2/39 UVEX6808.2/47
ESD Safety Shoes, Lace-Up, uvex 1 x-craft planet, 68082
Chaussures et bottes
The sporty uvex 1 X-craft safety shoe range takes light applications in trade, industry and logistics to the next level. The uvex 1 X-craft is designed for achievers and provides support for a wide variety of activities and working environments.
  • Sporty, especially light S1 PL safety shoe made from recycled and sustainable materials
  • Made from synthetic materials, so suitable for people allergic to chrome
  • All sole materials are free of silicones, plasticisers and other substances that interfere with wetting agents
  • Individually adjustable, elastic laces with quick release
  • Breathable distance mesh lining made from 18% bamboo and 52% recycled polyamide
  • Sole width: 25,4 mm
Upper material: uvex x-dry knit planet knitted upper
Lining: Distance-Mesh from 18% bamboo and 52% recycled polyamide
Anti penetration: textile, 4,5 mm nail (L)
Toe cap: xenova composite
Scuff cap: injected with PU midsole (SC)
Total flexibility and support are guaranteed thanks to the sole that features the uvex bionom x principle and an integrated flex zone in the forefoot area. The uvex 1 X-craft also takes abrasion resistance, protection and breathability to the next level thanks to the dynamic scuff cap, technological outer shoe materials and the perforated tongue design. The i-PUREnrj planet midsole ensures consistent energy return a Whether it’s in terms of sustainability, comfort, sportiness or innovation, the uvex 1 X-craft is simply the next level of safety shoe.
Light and breathable uvex X-dry knit planet textile outer shoe material. Shock-absorbing uvex i-PUREnrj planet midsole with energy return, made with 15% recycled granulate from production surpluses. Abrasion-resistant uvex X-tended grip planet TPU outsole made with 10% recycled granulate from production surpluses. Foamed polyurethane scuff cap. Foamed polyurethane heel basket. 100% metal-free protective toe cap. Flexible textile penetration resistance. ESD-certified.
Certifications: ESD; EN 61340-4-3 and EN 61340-5-1, EN ISO 20345:2022 S1 PL FO SR SC.