Imprimer
Stainless Steel Buckets with Spout
Fournisseur: Adelphi Pharma Hygiene Products
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BUS4015G BUL4010P BUL4012P BUL4015P BSL4015P BSL4010P BUS4010G
COLDBUS4015GEA 933 EUR
COLDBUS4015G COLDBUL4010P COLDBUL4012P COLDBUL4015P COLDBSL4015P COLDBSL4010P COLDBUS4010G
Stainless Steel Buckets with Spout
Godets
Hygienic, high-quality stainless steel buckets meet international pharma quality compliance regulations. Available in 304 grade stainless steel, these buckets are bright polished throughout, graduated in 1 L intervals, and come complete with a swing handle.
  • Buckets available as standard, off-the-shelf for delivery within days
  • Buckets in 304 grade stainless steel
  • Optional lids available to purchase separately
  • Exceptional average surface finish of 0,4 µm Ra
  • Can be cleaned with alcohol-based disinfectants, and in industrial dishwashers/autoclaves
  • Optional material grade, surface finish, and TSE certification available upon request
A range of optional modifications are available to meet every requirement, including bespoke etching of logos and other information. The complete range includes the following design options: buckets, heavy-duty buckets, and buckets with spout.