Imprimer
Centrifuge columns for affinity chromatography
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
89868 89869 89896 89897 89898 69707
PIER89868EA 121 EUR
PIER89868 PIER89869 PIER89896 PIER89897 PIER89898 PIER69707
Centrifuge columns for affinity chromatography
Filtres Filtres pour centrifugation
PP column, PE filter, ~30 µm pore size. Pierce™ centrifuge columns are made from low protein binding polypropylene for compatibility with protein purification, and fit into standard centrifuge tubes for use in any centrifuge (1,5, 2, 15 and 50 ml).
Pierce centrifuge columns are convenient tools for handling 40 µl to 10 ml of an affinity support for purification. They can be used in either spin or gravity-flow format. The spin format reduces sample processing time and makes column washing and elution more efficient. For gravity-flow procedures and when slow processing time is desired for binding steps with dilute samples, Column extenders can be used to increase the column reservoir capacity.