Imprimer
Filter discs with fused glass rim, borosilicate glass, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
252024104 252024207 252024301 252024404 252050105 252050208 252050302 252050405 252090109 252090203 252090306 252090409
511-5111EA 52.1 EUR
511-5111 511-5112 511-5113 511-5114 511-5121 511-5122 511-5123 511-5124 511-5131 511-5132 511-5133 511-5134
Filter discs with fused glass rim, borosilicate glass, DURAN®
Filters Filter Discs
These filter discs feature a fused glass rim.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • USP standard
Made from clear DURAN® borosilicate glass 3.3.