Imprimer
Cell culture inserts, Snapwell™
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3407 3801
734-1566EA 259 EUR
734-1566 734-1646
Cell culture inserts, Snapwell™
Microplaques Inserts pour plaques de cultures
A modified Transwell® permeable support containing a 12 mm Ø membrane supported by a detachable ring. Once cells are grown to confluence on the Snapwell™ insert, the ring can be placed in a vertical or horizontal diffusion chamber. Packaged in six well plates.
Informations de commande: Snapwell™ inserts are supplied preloaded into 6-well plates.