Imprimer
Optifit pipette tips, 10 - 1000 µl, extended, ST (10x96)
Fournisseur: Sartorius
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
LH-X791000 LH-X791001
BHITLH-X791000EA 103 EUR
BHITLH-X791000 BHITLH-X791001
Optifit pipette tips, 10 - 1000 µl, extended, ST (10x96)
Pointes de pipettes
The Optifit Extended Plus tips are designed to improve pipetting reach, making them ideal for use with deep and narrow labware. The tip design allows for more efficient retrieval of samples, down to the last drop. The longer length of these tips helps prevent contamination by ensuring that the pipette does not come into contact with the walls of labware or containers during use.
  • One of the longest 1000 μl tips on the market
  • Narrow and extended tip form to reach easier to the bottom of deep and/or narrow vessels
  • Comes as standard or pre-sterilized with or
En savoir plus
New Product
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable