Imprimer
Collagenase Type I
Marques : STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
07415 07416 100-0677
STMC07415EA 158 EUR
STMC07415 STMC07416 STMC100-0677
Collagenase Type I
Suppléments et additifs pour culture cellulaire
For digestion of native collagen fibrils.
  • Compatible with a variety of cell tissue types
Digest native collagen fibrils in connective tissues with Collagenase Type I. Sourced from Clostridium histolyticum, this enzymatic preparation contains the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin. Collagenase Type I has been used for the digestion of many human tissues and cell types, including the intestine (Barthel et al.), mammary glands (Huss and Kratz), prostate (Le et al.), endothelial cells (Ganguly et al.), and dorsal root ganglia (Dib-Hajj et al.).
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').