Imprimer
Elastase
Marques : STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
07454 07453 100-0682
STMC07454EA 1380 EUR
STMC07454 STMC07453 STMC100-0682
Elastase
Suppléments et additifs pour culture cellulaire
For digestion of native elastin.
  • Digest native elastin effectively
  • Use in combination with other proteinases for successful digestion of fibrous tissue
Digest native elastin in cell cultures with Elastase. This serine protease is frequently used in combination with other proteases such as collagenase and trypsin to digest fibrous tissue. Elastase consists of a single polypeptide chain of 240 amino acid residues and contains four disulfide bridges (Shotton and Hartley). It is synthesized as a zymogen (proelastase) and then converted to the active form by limited trypsin proteolysis at its N-terminal. Elastase preferentially cleaves the peptide bond of C-terminal neutral, non-aromatic amino acid residues (Schellenberger et al.).
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').