Imprimer
Collagenase Type III
Marques : STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
07422 07423 100-0679
STMC07422EA 177 EUR
STMC07422 STMC07423 STMC100-0679
Collagenase Type III
Suppléments et additifs pour culture cellulaire
For digestion of native collagen fibrils.
  • Use in a variety of cell and tissue types
Digest native collagen fibrils in connective tissues with Collagenase Type III, sourced from Clostridium histolyticum. Collagenase preparations typically contain the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin (Kessler and Yaron). While Collagenase Type III is lower in secondary proteolytic activity (caseinase, clostripain, and trypsin), it has similar collagenolytic activity compared to other collagenase types. Collagenase Type III has been used for the digestion of many tissues and cell types, including mammary glands (Rønnov-Jessen et al.), colon cancer stem cells (Prince et al.), and breast epithelial tumor cells (Chan et al.).
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').