Imprimer
PCR tubes, Corning® Thermowell® GOLD and Thermowell
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6530 6531 6571 3744 3745 3746 3747 3749 3750
734-1758EA 97.1 EUR
734-1758 734-1759 734-1776 732-5582 732-5583 732-5584 732-5585 732-5587 732-5588
PCR tubes, Corning® Thermowell® GOLD and Thermowell
Tubes Tubes PCR
Manufactured from ultra-pure, medical grade PP suitable for RT-PCR.
  • Available with flat caps for easy piercing and marking, or domed caps for efficient heat transfer
  • Ultra-thin walls of uniform thickness accelerate heat transfer for shorter cycle times and higher product yields
  • 100% tested for well integrity
  • Certified DNase- and RNase-free and endotoxin-safe on a lot-by-lot basis
  • Autoclavable at 121 °C
  • Able to withstand centrifugation to 10000×g
Tubes are designed with longer cap flanges and sealed with a full-length insertion cap to safeguard against evaporation and to prevent accidental cap opening during thermal cycling.
Informations de livraison: Assorted pack includes the following colours: blue, green, orange, red, violet, and yellow.