Imprimer
Mesenchymal Stem Cells, BM, Healthy
Marques : PROVITRO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
12196102005 12296102010
PRTV12196102005EA 1210 EUR
PRTV12196102005 PRTV12296102010
Mesenchymal Stem Cells, BM, Healthy
Cellules
Bone Marrow CD34⁺ cells, enriched with CD34⁺ markers, are pivotal for haematopoiesis and immune modulation. These unique cells play a crucial role in hematopoietic stem cell transplantation and immune reconstitution. They contribute to the formation of various blood cell lineages, including immune effectors such as natural killer cells and cytotoxic T-cells, shaping both the adaptive and functional aspects of the immune system.
  • Purity >90% by flow cytometry
  • Test negative for Hepatitis B and C, HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV
  • Cryopreserved in CryoStor® CS10 media
Cells were collected from from a healthy human donors using permission forms and procedures that have been approved by the Institutional Review Board (IRB).
Storage: For prolonged cell preservation or resuscitation, it is recommended to prepare the cells for extended storage in the vapor phase of liquid nitrogen. Storage of cells in the liquid phase of nitrogen is strongly discouraged to prevent potential cross-contamination risks when removing from storage. Long-term storage under −175 °C and for short-term storage, about one or two weeks at −80 °C is permitted, but it is recommended to minimise this duration to ensure optimal stability. Thawed samples should be used immediately and not stored.