Imprimer
PBMC, Healthy, ImmunoSafe-HLA1 and HLA2
Marques : PROVITRO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
12194113010 12194114010
PRTV12194113010EA 4000 EUR
PRTV12194113010 PRTV12194114010
PBMC, Healthy, ImmunoSafe-HLA1 and HLA2
Cellules
Our ImmunoSafe-HLA I and ImmunoSafe-HLA II panels are meticulously designed to enhance the predictivity of your preclinical assays. With 10 HLA Class I- or HLA Class II-matched donors, these panels can be tailored to suit your specific assay requirements. By providing a comprehensive resource to mimic diverse immunological responses, they empower researchers to achieve more accurate and relevant experimental outcomes.
  • Purity >90% by flow cytometry
  • Test negative for Hepatitis B and C, HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV
  • Cryopreserved in CryoStor® CS10 media
Their versatility and adaptability ensure seamless integration into existing research protocols, making them invaluable tools for deciphering the complexities of the immune system and driving groundbreaking discoveries in biomedical research.
Cells were collected from from a healthy human donors using permission forms and procedures that have been approved by the Institutional Review Board (IRB).
Storage: For prolonged cell preservation or resuscitation, it is recommended to prepare the cells for extended storage in the vapor phase of liquid nitrogen. Storage of cells in the liquid phase of nitrogen is strongly discouraged to prevent potential cross-contamination risks when removing from storage. Long-term storage under −175 °C and for short-term storage, about one or two weeks at −80 °C is permitted, but it is recommended to minimise this duration to ensure optimal stability. Thawed samples should be used immediately and not stored.