Imprimer
Drawer Racks, Stainless Steel
Fournisseur: RATIOLAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5400046 5400019 5400050 5400047
RATI5400046EA 215 EUR
RATI5400046 RATI5400019 RATI5400050 RATI5400047
Drawer Racks, Stainless Steel
Portoirs Portoirs pour cryoboîtes
Stainless steel racks for storing the cryo boxes in freezer cabinets and chests.
  • Cabinet slide-ins without pull-out stop
  • Easy-to-clean polished surface
  • Robust design
Racks for for all cryo boxes format 130×130; 133×133 and 136×136 mm.