Imprimer
EasySep™ Mouse PE Positive Selection Kit II
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17666 17696 17666RF 17696RF
STMC17666EA 1130 EUR
STMC17666 STMC17696 STMC17666RF STMC17696RF
EasySep™ Mouse PE Positive Selection Kit II
Milieux de séparation cellulaire
Immunomagnetic positive selection of mouse cells labelled with PE-conjugated antibodies.
  • Fast and easy-to-use
  • No columns required
The EasySep™ Mouse PE Positive Selection Kit II is designed to isolate cells that are labelled with PE-conjugated antibodies. Desired cells are targeted with antibody complexes recognising PE and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This kit replaces the EasySep™ Mouse PE Positive Selection Kit.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').