Imprimer
EasySep™ Human CD33 Positive Selection Kit II
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17876RF 17876
STMC17876RFEA 1410 EUR
STMC17876RF STMC17876
EasySep™ Human CD33 Positive Selection Kit II
Milieux de séparation cellulaire
Immunomagnetic positive selection of human CD33+ cells (e.g. myeloid cells).
  • Fast and easy-to-use
  • Up to 98% purity
  • No columns required
  • Compatible across EasySep™, 'The Big Easy', and RoboSep™-S platforms
The EasySep™ Human CD33 Positive Selection Kit II is designed to isolate CD33+ cells from ammonium chloride lysed blood or bone marrow, or from nucleated cells prepared by density separation with RosetteSep™ DM-M density medium by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD33 and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD33 antigen is expressed on cells of the myeloid lineage.
This product replaces the EasySep™ Human CD33 Positive Selection Kit for even faster cell isolations.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').