Imprimer
EasySep™ Human CD138 Positive Selection Kit II
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17877 17877RF
STMC17877EA 1250 EUR
STMC17877 STMC17877RF
EasySep™ Human CD138 Positive Selection Kit II
Milieux de séparation cellulaire
Immunomagnetic positive selection of human CD138+ cells (e.g. normal or malignant plasma cells for multiple myeloma research).
  • Fast and easy-to-use
  • No columns required
The EasySep™ Human CD138 Positive Selection Kit II is designed to isolate CD138+ to learn how to process bone marrow samples and isolate CD138+ plasma cells to increase sensitivity in FISH testing for multiple myeloma. You can also browse our Frequently Asked Questions about isolating CD138+ plasma cells to improve FISH sensitivity.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').