Imprimer
EasySep™ Human Resting CD4+ T Cell Isolation Kits
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17962RF 17962
STMC17962RFEA 2040 EUR
STMC17962RF STMC17962
EasySep™ Human Resting CD4+ T Cell Isolation Kits
Milieux de séparation cellulaire
Immunomagnetic isolation of untouched human resting CD4+ T cells.
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Isolated cells are untouched
The EasySep™ Human Resting CD4+ T Cell Isolation Kit is designed to isolate resting CD4+ T cells from fresh or previously frozen human peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep™ procedure involves labelling unwanted cells with antibody complexes and magnetic particles. The magnetically labelled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').