Imprimer
EasySep™ Human B Cell Enrichment Kit II Without CD43 Depletion
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17963 17963RF
STMC17963EA 1810 EUR
STMC17963 STMC17963RF
EasySep™ Human B Cell Enrichment Kit II Without CD43 Depletion
Milieux de séparation cellulaire
Immunomagnetic isolation of untouched human B cells without CD43 depletion.
  • Fast and easy-to-use
  • Up to 99% purity
  • Isolated cells are untouched
The EasySep™ Human B Cell Enrichment Kit II without CD43 Depletion is designed to isolate B cells by negative selection from fresh or previously frozen PBMCs of individuals with B cell leukaemia or lymphoma, or other disease states in which B cells may express CD43. The EasySep™ procedure involves labelling unwanted cells with antibody complexes and magnetic particles. The magnetically labelled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
This kit replaces the EasySep™ Human B Cell Enrichment Kit without CD43 for even faster cell isolations.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').