Imprimer
EasySep™ Non-Human Primate Custom Positive Selection Kits
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
18809 18809RF
STMC18809EA 0 EUR
STMC18809 STMC18809RF
EasySep™ Non-Human Primate Custom Positive Selection Kits
Milieux de séparation cellulaire
Immunomagnetic positive selection of non-human primate cells.
The EasySep™ Non-Human Primate Custom Positive Selection Kit is designed to isolate any cell type of interest from fresh or previously frozen peripheral blood mononuclear cells by positive selection. STEMCELL Technologies offers a wide selection of antibodies for custom separation and will work with you to isolate a cell type of interest. Please contact our technical support team for more information.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form,En savoir plus