Imprimer
EasySep™ Mouse CD11b Positive Selection Kit II
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
18970 18970RF
STMC18970EA 1130 EUR
STMC18970 STMC18970RF
EasySep™ Mouse CD11b Positive Selection Kit II
Milieux de séparation cellulaire
Immunomagnetic positive selection of CD11b+ cells (e.g. monocytes) from mouse splenocytes or other tissues.
  • Fast, easy-to-use, and column-free
  • Up to 95% purity (for bone marrow, purity can be up to 99%)
  • Isolated cells are not fluorochrome-labelled
The EasySep™ Mouse CD11b Positive Selection Kit II isolates highly purified CD11b+ cells from mouse splenocytes, bone marrow, lungs, brains or other tissues by immunomagnetic positive selection. Desired cells are targeted with antibodies and magnetic particles, and isolated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Isolated cells are immediately ready for downstream applications such as flow cytometry, culture, and cell-based experiments.
This product replaces the EasySep™ Mouse CD11b Positive Selection Kit for even faster cell isolations and does not result in the labelling of isolated cells with PE.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').