Imprimer
EasySep™ Human CD4+ T Cell Enrichment Kits
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
19052 19052RF
STMC19052EA 1810 EUR
STMC19052 STMC19052RF
EasySep™ Human CD4+ T Cell Enrichment Kits
Milieux de séparation cellulaire
Immunomagnetic isolation of untouched human CD4+ T cells.
  • Fast, easy-to-use and column-free
  • Up to 97% purity
  • Untouched, viable cells
The EasySep™ Human CD4+ T Cell Enrichment Kit is designed to isolate CD4+ T cells from fresh or previously frozen peripheral mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-CD4+ T cells and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
For even faster cell isolations, we recommend the new EasySep™ Human CD4+ T Cell Isolation Kit which isolates cells in just 8 minutes.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').