Imprimer
EasySep™ Human NK Cell Enrichment Kits
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
19055 19055RF
STMC19055EA 1880 EUR
STMC19055 STMC19055RF
EasySep™ Human NK Cell Enrichment Kits
Milieux de séparation cellulaire
Immunomagnetic isolation of untouched human NK cells.
  • Fast, easy-to-use and column-free
  • Up to 95% purity
  • Untouched, viable cells
The EasySep™ Human NK Cell Enrichment Kit is designed to isolate NK cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-NK cells and dextran-coated magnetic particles. The labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
For even faster cell isolations, we recommend the new EasySep™ Human NK Cell Isolation Kit which isolates cells in just 8 minutes.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').