Imprimer
EasySep™ Human Monocyte Enrichment Kits without CD16 Depletion
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
19058 19058RF 100-1525
STMC19058EA 1810 EUR
STMC19058 STMC19058RF STMC100-1525
EasySep™ Human Monocyte Enrichment Kits without CD16 Depletion
Milieux de séparation cellulaire
Immunomagnetic isolation of untouched human monocytes
  • Fast, easy-to-use, and column-free
  • Up to 90% purity
  • Untouched, viable cells
The EasySep™ Human Monocyte Enrichment Kit without CD16 Depletion is designed to isolate monocytes and CD14+CD16+ monocytes from fresh or previously frozen peripheral blood mononuclear cells by negative selection. The CD14+CD16+ subset of monocytes (Catalog #19359), which contains anti-CD16.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').