Imprimer
EasySep™ Human Plasmacytoid DC Enrichment Kits
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
19062 19062RF
STMC19062EA 2150 EUR
STMC19062 STMC19062RF
EasySep™ Human Plasmacytoid DC Enrichment Kits
Milieux de séparation cellulaire
Immunomagnetic isolation of untouched human plasmacytoid dendritic cells (pDC)
  • Fast, easy-to-use and column-free
  • Up to 97% purity
  • Untouched, viable cells
The EasySep™ Human Plasmacytoid DC Enrichment Kit is designed to isolate plasmacytoid dendritic cells (pDC) from fresh peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-pDCs and dextran-coated magnetic particles. The labelled cells are separated using the EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').