Imprimer
EasySep™ Human Gamma/Delta T Cell Isolation Kits
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
19255 19255RF
STMC19255EA 1880 EUR
STMC19255 STMC19255RF
EasySep™ Human Gamma/Delta T Cell Isolation Kits
Milieux de séparation cellulaire
Immunomagnetic isolation of untouched human gamma/delta T cells
  • Fast, easy-to-use and column-free
  • Up to 97% purity
  • Isolated cells are untouched
The EasySep™ Human Gamma/Delta T Cell Isolation Kit is designed to isolate gamma/delta T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-gamma/delta T cells and dextran-coated magnetic particles. The labelled cells are separated using the EasySep™ magnet without the use of columns. The desired cells are poured off into a new tube. To obtain a custom kit optimized for culture-expanded PBMCs, contact STEMCELL Technologies’ Technical Support.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').