Imprimer
EasySep™ Human Custom Enrichment Kits
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
19309 19309RF
STMC19309EA 0 EUR
STMC19309 STMC19309RF
EasySep™ Human Custom Enrichment Kits
Milieux de séparation cellulaire
Immunomagnetic isolation of untouched human cells
The EasySep™ Human Custom Enrichment Kit is designed to isolate any cell type of interest from fresh or previously frozen peripheral blood mononuclear cells by negative selection. STEMCELL Technologies offers a wide selection of antibodies for custom separation and will work with you to isolate a cell type of interest. Please contact our technical support team for more information.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').