Imprimer
EasySep™ Human Naïve CD4+ T Cell Isolation Kits
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
19555 19555RF
STMC19555EA 1810 EUR
STMC19555 STMC19555RF
EasySep™ Human Naïve CD4+ T Cell Isolation Kits
Milieux de séparation cellulaire
Immunomagnetic negative selection of untouched human naïve CD4+ T cells
  • Fast, easy-to-use and column-free
  • Up to 96% purity
  • Isolated cells are untouched
The EasySep™ Human Naïve CD4+ T Cell Isolation Kit is designed to isolate naive CD4+ T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes and dextran-coated magnetic particles. CD45RO+ cells are targeted for removal with a biotinylated anti-CD45RO antibody, and a bispecific Tetrameric Antibody Complex that recognizes biotin and dextran. The labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
For even faster cell isolations, we recommend the new EasySep™ Human Naïve CD4+ T Cell Isolation Kit II which isolates cells in just 11 minutes.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').