Imprimer
EasySep™ Direct Human T Cell Isolation Kits
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
19661 19661RF
STMC19661EA 1150 EUR
STMC19661 STMC19661RF
EasySep™ Direct Human T Cell Isolation Kits
Milieux de séparation cellulaire
Immunomagnetic isolation of T cells directly from whole blood
  • >99,9% RBC depletion without the need for density gradient centrifugation, sedimentation, or lysis
  • Up to 97% purity of isolated cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
This cell separation kit can be used to isolate highly purified T cells directly from human whole blood by immunomagnetic negative selection.
This kit targets non-T cells for removal with antibodies recognising specific surface markers. Unwanted cells are labelled with antibodies and EasySep™En savoir plus