Imprimer
EasySep™ Direct Human CD4+ T Cell Isolation Kits
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
19662 19662RF
STMC19662EA 1400 EUR
STMC19662 STMC19662RF
EasySep™ Direct Human CD4+ T Cell Isolation Kits
Milieux de séparation cellulaire
Immunomagnetic isolation of CD4+ T cells directly from whole blood
  • >99,9% RBC depletion without the need for density gradient centrifugation, sedimentation or lysis
  • Up to 96% purity of isolated cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
Isolate CD4+ T cells directly from human whole blood by immunomagnetic negative selection. Desired cells undergo minimal manipulation and are untouched and highly purified.
This product labels RBCs, platelets and unwanted cells with antibody complexes and magnetic particles. TheEn savoir plus
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable