Imprimer
EasySep™ Mouse Pan-Naïve T Cell Isolation Kits
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
19848 19848RF
STMC19848EA 1370 EUR
STMC19848 STMC19848RF
EasySep™ Mouse Pan-Naïve T Cell Isolation Kits
Milieux de séparation cellulaire
Immunomagnetic isolation of untouched mouse pan-naïve T cells
  • Fast and easy-to-use
  • Up to 97% purity
  • No columns required
  • Untouched, viable cells
The EasySep™ Mouse Pan-Naïve T Cell Isolation Kit is designed to isolate T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-naïve T cells to learn about our next-generation EasySep™ mouse cell isolation kits, featuring RapidSphere™ technology.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').